This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01173919_m1
See other YAP1 GE Assays ›
Gene Symbol:
Yap1
Gene Name:
yes-associated protein 1
Gene Aliases:
AI325207, Yap, Yap65, Yki, Yorkie
Chromosome Location:
Chr.9: 7931999 - 8004596 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01173919_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Yap1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001171147.1 NP_001164618.1 5 - 6 1146 74
GenBank mRNA AK204312.1 - 1 - 2 184 74
BC014733.1 - 5 - 6 1143 4239820 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900001724.mm.1

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

transcription cofactor

Gene Ontology Categories:

Function(s) Process(es)

cell morphogenesis
vasculogenesis
embryonic heart tube morphogenesis
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
cellular response to DNA damage stimulus
cell proliferation
positive regulation of cell proliferation
regulation of keratinocyte proliferation
keratinocyte differentiation
negative regulation of epithelial cell differentiation
notochord development
somatic stem cell population maintenance
hippo signaling
regulation of cell proliferation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of organ growth
paraxial mesoderm development
lateral mesoderm development
regulation of neurogenesis
contact inhibition
bud elongation involved in lung branching
lung epithelial cell differentiation
regulation of canonical Wnt signaling pathway
cellular response to retinoic acid
cellular response to gamma radiation
regulation of stem cell proliferation
regulation of metanephric nephron tubule epithelial cell differentiation
positive regulation of canonical Wnt signaling pathway
positive regulation of stem cell population maintenance
negative regulation of nucleic acid-templated transcription
negative regulation of stem cell differentiation
negative regulation of extrinsic apoptotic signaling pathway
RNA polymerase II core promoter sequence-specific DNA binding
transcription factor activity, RNA polymerase II transcription factor binding
chromatin binding
transcription coactivator activity
transcription corepressor activity
protein binding
protein C-terminus binding
transcription regulatory region DNA binding
proline-rich region binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0