This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01174359_m1
See other NXPE2 GE Assays ›
Gene Symbol:
Nxpe2
Gene Name:
neurexophilin and PC-esterase domain family, member 2
Gene Aliases:
4432416J03Rik, AI323362, Fam55b
Chromosome Location:
Chr.9: 48318003 - 48353453 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01174359_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nxpe2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK157096.1 - 1 - 2 271 66
BC140955.1 - 1 - 2 216 9055567 66

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK157096.1, BC140955.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900018628.mm.1 NM_030069.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900018628.mm.1 NM_030069.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900018628.mm.1 TC0900002233.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900018629.mm.1 NM_030069.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900018629.mm.1 NM_030069.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900018629.mm.1 TC0900002233.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-f5rhb:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0