This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01174364_m1
See other CADM1 GE Assays ›
Gene Symbol:
Cadm1
Gene Name:
cell adhesion molecule 1
Gene Aliases:
2900073G06Rik, 3100001I08Rik, AI987920, Bl2, Igsf4, Igsf4a, Necl2, RA175, RA175A, RA175B, RA175C, RA175N, ST17, SgIGSF, SynCam, Tslc1
Chromosome Location:
Chr.9: 47530239 - 47853385 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01174364_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cadm1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_207675.2 NP_997558.2 7 - 8 1018 136
GenBank mRNA AB064265.2 - 7 - 8 997 136
AK013911.1 - 7 - 8 1112 136
EU541476.1 - 1 - 2 167 136
JQ782219.1 - 7 - 8 997 136

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900000573.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cadm1

Gene Ontology Categories:

Function(s) Process(es)

liver development
immune system process
apoptotic process
cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
multicellular organism development
spermatogenesis
synapse assembly
cell recognition
unidimensional cell growth
calcium-independent cell-cell adhesion via plasma membrane cell-adhesion molecules
cell differentiation
susceptibility to natural killer cell mediated cytotoxicity
positive regulation of natural killer cell mediated cytotoxicity
positive regulation of cytokine secretion
detection of stimulus
bone development
receptor binding
protein binding
PDZ domain binding
protein homodimerization activity
cell adhesion molecule binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900004052.mm.1 NM_018770.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900004052.mm.1 NM_018770.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900004052.mm.1 NM_207675.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900004052.mm.1 NM_207675.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900004052.mm.1 TC0900000573.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0