This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01174694_m1
See other HMGCLL1 GE Assays ›
Gene Symbol:
Hmgcll1
Gene Name:
3-hydroxymethyl-3-methylglutaryl-Coenzyme A lyase-like 1
Gene Aliases:
BC037381
Chromosome Location:
Chr.9: 76014650 - 76136350 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01174694_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hmgcll1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_173731.2 NP_776092.1 4 - 5 492 101
GenBank mRNA AK141622.1 - 4 - 5 455 101
AK164164.1 - 4 - 5 455 101
BC037381.1 - 4 - 5 492 5368383 101

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900001022.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hmgcll1

Panther Classification:

Molecular Function -

lyase

Gene Ontology Categories:

Function(s) Process(es)

ketone body biosynthetic process
catalytic activity
hydroxymethylglutaryl-CoA lyase activity
lyase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900008085.mm.1 NM_173731.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900008085.mm.1 NM_173731.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900008085.mm.1 TC0900001022.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-tks2p:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0