This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01174817_m1
See other FAT3 GE Assays ›
Gene Symbol:
Fat3
Gene Name:
FAT tumor suppressor homolog 3 (Drosophila)
Gene Aliases:
9430076A06Rik, D430038H04Rik, Gm1132, Gm510, MFAT3F
Chromosome Location:
Chr.9: 15910189 - 16502879 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01174817_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fat3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001080814.1 NP_001074283.1 16 - 17 10357 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900001792.mm.1

Set Membership:

Amplicon length between 71 and 85 Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

cell morphogenesis involved in differentiation
neuron migration
cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
multicellular organism development
retina layer formation
negative regulation of dendrite development
calcium ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900015102.mm.1 NM_001080814.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900015102.mm.1 NM_001080814.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900015102.mm.1 TC0900001792.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0