This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01174906_m1
See other SDHD GE Assays ›
Gene Symbol:
Sdhd
Gene Name:
succinate dehydrogenase complex, subunit D, integral membrane protein
Gene Aliases:
3110001M13Rik, AVLL5809, C78570, PRO19626
Chromosome Location:
Chr.9: 50596340 - 50603849 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01174906_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sdhd
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025848.3 NP_080124.1 2 - 3 232 63
GenBank mRNA AK013503.1 - 2 - 3 197 63
AK135970.1 - 2 - 3 192 63
BC145731.1 - 2 - 3 192 40131034 63
BC145733.1 - 2 - 3 192 40131036 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900002271.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sdhd

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

tricarboxylic acid cycle
transport
regulation of catecholamine secretion
oxidation-reduction process
cellular response to hypoxia
succinate dehydrogenase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900018883.mm.1 NM_025848.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900018883.mm.1 NM_025848.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900018883.mm.1 TC0900002271.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0