This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01174944_m1
See other HACD3 GE Assays ›
Gene Symbol:
Hacd3
Gene Name:
3-hydroxyacyl-CoA dehydratase 3
Gene Aliases:
4930523M17Rik, AW742319, B-ind1, Hcad3, Hspc121, Ptplad1
Chromosome Location:
Chr.9: 64986983 - 65021735 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01174944_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hacd3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021345.2 NP_067320.2 1 - 2 260 73
GenBank mRNA AK050556.1 - 1 - 2 254 73
AK051735.1 - 1 - 2 219 73
AK052865.1 - 1 - 2 257 73
AK159494.1 - 1 - 2 226 73
BC031755.1 - 1 - 2 244 4488391 73
CJ176979.1 - 1 - 2 260 73
Z97207.2 - 1 - 2 86 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900002503.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hacd3

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
fatty acid metabolic process
fatty acid biosynthetic process
I-kappaB kinase/NF-kappaB signaling
JNK cascade
Rho protein signal transduction
fatty acid elongation
very long-chain fatty acid biosynthetic process
positive regulation of viral genome replication
positive regulation by virus of viral protein levels in host cell
protein binding
lyase activity
3-hydroxyacyl-CoA dehydratase activity
enzyme binding

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 828518 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0