This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01175022_g1
See other RBPMS2 GE Assays ›
Gene Symbol:
Rbpms2
Gene Name:
RNA binding protein with multiple splicing 2
Gene Aliases:
2400008B06Rik, AI316523
Chromosome Location:
Chr.9: 65629336 - 65660536 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01175022_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rbpms2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028030.3 NP_082306.2 3 - 4 435 66
GenBank mRNA AK010301.1 - 4 - 5 387 66
AK144659.1 - 3 - 4 460 66
AK156633.1 - 3 - 4 435 66
BC021788.1 - 3 - 4 433 4236661 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900000848.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rbpms2

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

biological_process
nucleotide binding
molecular_function
nucleic acid binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900006503.mm.1 NM_028030.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900006503.mm.1 NM_028030.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900006503.mm.1 TC0900000848.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900006502.mm.1 NM_028030.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900006502.mm.1 NM_028030.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900006502.mm.1 TC0900000848.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline