This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01175029_m1
See other IGDCC4 GE Assays ›
Gene Symbol:
Igdcc4
Gene Name:
immunoglobulin superfamily, DCC subclass, member 4
Gene Aliases:
DDM36, Nope
Chromosome Location:
Chr.9: 65101461 - 65137947 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01175029_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Igdcc4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001290315.1 NP_001277244.1 3 - 4 610 104
NM_020043.3 NP_064427.2 3 - 4 610 104
GenBank mRNA AB052620.1 - 3 - 4 610 104
AB052621.1 - 3 - 4 610 104
AK079073.1 - 3 - 4 575 104
AK122535.1 - 539 104
AK143256.1 - 3 - 4 562 103
BB624536.1 - 3 - 4 601 104
CD352400.1 - 2 - 3 247 104

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Igdcc4

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline