This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01175341_m1
See other EIF3K GE Assays ›
Gene Symbol:
Eif3k
Gene Name:
eukaryotic translation initiation factor 3, subunit K
Gene Aliases:
1200009C21Rik, Eif3s12
Chromosome Location:
Chr.7: 28971372 - 28981889 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01175341_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eif3k
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001285942.1 NP_001272871.1 6 - 7 677 61
NM_028659.3 NP_082935.1 7 - 8 776 61
GenBank mRNA AK004664.1 - 7 - 8 703 61
BC027638.1 - 6 - 7 576 1348838 61
BC091749.1 - 7 - 8 680 3661337 61
CD802664.1 - 1 - 2 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700002710.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eif3k

Panther Classification:

Molecular Function -

translation factor translation initiation factor translational protein

Gene Ontology Categories:

Function(s) Process(es)

translation
translational initiation
regulation of translational initiation
RNA binding
translation initiation factor activity
ribosome binding

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25024699 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25024698 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2016387 BC027638.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2016387 BC091749.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2016387 NM_028659.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2016387 AK004664.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P341025 BC091749.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P341025 NM_028659.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P341025 AK004664.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P341025 AK037834.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P341025 BC091749.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P341025 NM_028659.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P341025 AK004664.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P341025 AK037834.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2016388 BC027638.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2016388 BC091749.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2016388 NM_028659.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2016388 AK004664.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2016388 AK158875.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 515556 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 25612 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 924682 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 62498 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 69008 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 149430 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P146864 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P138997 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2016387 BC027638.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2016387 BC091749.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2016387 NM_028659.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2016387 AK004664.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2016388 BC027638.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2016388 BC091749.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2016388 NM_028659.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2016388 AK004664.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2016388 AK158875.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P341025 BC091749.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P341025 NM_028659.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P341025 AK004664.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P341025 AK037834.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P341025 BC091749.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P341025 NM_028659.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P341025 AK004664.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P341025 AK037834.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P341025 BC091749.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P341025 NM_028659.2 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P341025 AK004664.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P341025 AK037834.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P341025 BC091749.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P341025 NM_028659.2 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P341025 AK004664.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P341025 AK037834.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25024678 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25024694 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25024696 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25024675 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25024708 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25024690 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25024701 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25024695 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25024689 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25024697 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25024703 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25024682 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25024686 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25024699 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25024677 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25024706 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25024683 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25024704 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25024709 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25024680 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25024700 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25024692 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P138997 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P146864 - -
Whole Mouse Genome G4122A A_51_P341025 BC091749.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P341025 NM_028659.2 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P341025 AK004664.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P341025 AK037834.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P341025 BC091749.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P341025 NM_028659.2 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P341025 AK004664.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P341025 AK037834.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109822 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102974 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P138997 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P146864 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06587232 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06587238 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06348800 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06587220 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06348803 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06587250 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06587227 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25024698 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25024699 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024697 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024595 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024628 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024656 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024640 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024735 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024683 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024732 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024673 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024752 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024738 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024672 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024740 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024626 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024631 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024718 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024614 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024604 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024761 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024693 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024644 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024695 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024689 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024633 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024727 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024601 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024677 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024745 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024589 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024730 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024663 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024616 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024645 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024664 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024701 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024709 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024686 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024658 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024708 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024666 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024756 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024653 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024728 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024699 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024643 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024602 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024621 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024711 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024720 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024668 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024692 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024713 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024639 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024680 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024758 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024651 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024743 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024611 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024703 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024674 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024716 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024637 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024721 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024607 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024670 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024594 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024660 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024706 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024646 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024615 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024624 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024635 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024760 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024750 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024690 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024710 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024590 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024694 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25024746 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06368597 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06045091 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05936117 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25024703 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25024695 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25024683 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25024686 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25024677 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25024701 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25024689 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05936116 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05982142 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25024660 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06513338 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25024680 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25024685 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25024692 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25024678 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06045090 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25024693 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25024690 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25024668 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25024666 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05954635 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25024688 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06348798 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25024694 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06348807 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25024699 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25024704 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05917853 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10561558 - -
Affymetrix Mouse Gene 1.0 ST Array 10561554 - -
Affymetrix Mouse Gene 1.0 ST Array 10561553 - -
Affymetrix Mouse Gene 1.0 ST Array 10561556 - -
Affymetrix Mouse Gene 1.0 ST Array 10561559 - -
Affymetrix Mouse Gene 1.0 ST Array 10561555 - -
Affymetrix Mouse Gene 1.0 ST Array 10561560 - -
Affymetrix Mouse Gene 1.0 ST Array 10561557 - -
Affymetrix HT MG-430 PM Array Plate 1452674_PM_a_at - -
Affymetrix Mouse Gene 1.1 ST Array 10561552 - -
Mouse Genome 430 2.0 Array 1452674_a_at - -
Mouse Genome 430 2.0 Array 1452674_PM_a_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 73830_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 73830_at - -
Affymetrix Mouse Clariom S Array PSR0700023783.mm.1 TC0700002710.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700023783.mm.1 NM_001285942.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700023783.mm.1 NM_028659.3 Refseq
Mouse Expression Set 430 1452674_a_at - -
Mouse Expression Set 430 1452674_PM_a_at - -
Murine Genome U74v2 Set 95715_PM_at - -
Murine Genome U74v2 Set 95715_at - -
Mouse Genome 430A 2.0 Array 1452674_a_at - -
Mouse Genome 430A 2.0 Array 1452674_PM_a_at - -
Affymetrix HT Mouse Genome 430A Array 1452674_a_at - -
Affymetrix HT Mouse Genome 430A Array 1452674_PM_a_at - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 100780538 - -
Illumina mouse-6 v1.0 expression beadchip 5720372 - -
Illumina mouseRef-8 v1.0 expression beadchip 0005720372 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2718416 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1226498 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2718416 - -
mouse-6 v1.1 (Illumina) scl31564.8.5_30-S - -
Illumina Sentrix mouse-6.v1 bead array 100780538 - -
Illumina Sentrix mouse-6.v1 bead array 5720372 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl31564.8.5_30-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 5720372 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 100780538 - -
MouseRef-8 v2 Expression BeadChip ILMN_2718416 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2718416 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1226498 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl31564.8.5_30-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0