This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01175859_g1
See other ICAM5 GE Assays ›
Gene Symbol:
Icam5
Gene Name:
intercellular adhesion molecule 5, telencephalin
Gene Aliases:
CD50, ICAM-3, Icam3, TLN, Tlcn
Chromosome Location:
Chr.9: 21032038 - 21039036 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01175859_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Icam5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008319.2 NP_032345.2 9 - 10 2321 106
GenBank mRNA AK135040.1 - 9 - 10 2234 106
BC113140.1 - 9 - 10 2269 40048010 106
U06483.1 - 9 - 10 2269 106

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900000186.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Icam5

Gene Ontology Categories:

Function(s) Process(es)

phagocytosis
cell adhesion
single organismal cell-cell adhesion
integrin binding
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900001233.mm.1 NM_008319.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900001233.mm.1 NM_008319.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900001233.mm.1 TC0900000186.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline