This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01176004_g1
See other PPA1 GE Assays ›
Gene Symbol:
Ppa1
Gene Name:
pyrophosphatase (inorganic) 1
Gene Aliases:
2010317E03Rik, Pyp
Chromosome Location:
Chr.10: 61648621 - 61674165 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01176004_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppa1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026438.4 NP_080714.2 7 - 8 745 147
GenBank mRNA AK008575.1 - 7 - 8 746 147
AK088397.1 - 7 - 8 741 147
AK145640.1 - 7 - 8 795 147
AK146300.1 - 7 - 8 708 147
AK151512.1 - 7 - 8 736 147
BC010468.1 - 7 - 8 679 3585780 147
CT010203.1 - 7 - 8 638 147

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000000571.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppa1

Panther Classification:

Molecular Function -

pyrophosphatase

Gene Ontology Categories:

Function(s) Process(es)

phosphate-containing compound metabolic process
magnesium ion binding
inorganic diphosphatase activity
pyrophosphatase activity
hydrolase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000003865.mm.1 NM_026438.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000003865.mm.1 NM_026438.4 Refseq
Affymetrix Mouse Clariom S Array PSR1000003865.mm.1 TC1000000571.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline