This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01176005_m1
See other PPA1 GE Assays ›
Gene Symbol:
Ppa1
Gene Name:
pyrophosphatase (inorganic) 1
Gene Aliases:
2010317E03Rik, Pyp
Chromosome Location:
Chr.10: 61648621 - 61674165 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01176005_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppa1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026438.4 NP_080714.2 8 - 9 833 108
GenBank mRNA AK008575.1 - 8 - 9 834 108
AK020291.1 - 8 - 9 849 108
AK088397.1 - 8 - 9 829 108
AK145640.1 - 8 - 9 883 108
AK146300.1 - 8 - 9 796 108
AK151512.1 - 8 - 9 824 108
AK201001.1 - 2 - 3 108
AK216005.1 - 2 - 3 187 108
BC010468.1 - 8 - 9 767 3585780 108
CT010203.1 - 8 - 9 726 108

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000000571.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppa1

Panther Classification:

Molecular Function -

pyrophosphatase

Gene Ontology Categories:

Function(s) Process(es)

phosphate-containing compound metabolic process
magnesium ion binding
inorganic diphosphatase activity
pyrophosphatase activity
hydrolase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000003866.mm.1 NM_026438.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000003866.mm.1 NM_026438.4 Refseq
Affymetrix Mouse Clariom S Array PSR1000003866.mm.1 TC1000000571.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline