Assay Details
| Gene Symbol: Nfrkb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172766.3 | NP_766354.2 | 9 - 10 | 1197 | 58 | |
| GenBank mRNA | AK041206.1 | - | 9 - 10 | 1435 | 58 | |
| BC033595.1 | - | 9 - 10 | 1187 | 4947357 | 58 | |
| BC059861.1 | - | 9 - 10 | 1244 | 58 | ||
| BC059900.1 | - | 1 - 2 | 109 | 58 | ||
| BC060076.1 | - | 9 - 10 | 1426 | 58 | ||
| CB244971.1 | - | 5 - 6 | 557 | 58 | ||
More Information
Set Membership: |
|

Nfrkb
Genomic Map