This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01176319_m1
See other PPP1R14A GE Assays ›
Gene Symbol:
Ppp1r14a
Gene Name:
protein phosphatase 1, regulatory (inhibitor) subunit 14A
Gene Aliases:
1110001M11Rik, Cpi17
Chromosome Location:
Chr.7: 29289320 - 29293390 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01176319_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp1r14a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026731.3 NP_081007.2 1 - 2 216 83
GenBank mRNA AF352573.1 - 1 - 2 224 83
AY050672.1 - 1 - 2 215 83
BC010832.1 - 1 - 2 216 4216018 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700000527.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppp1r14a

Panther Classification:

Molecular Function -

phosphatase modulator

Gene Ontology Categories:

Function(s) Process(es)

regulation of phosphorylation
protein phosphatase inhibitor activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700004488.mm.1 TC0700000527.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700004488.mm.1 NM_026731.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700004488.mm.1 NM_026731.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700004489.mm.1 TC0700000527.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700004489.mm.1 NM_026731.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700004489.mm.1 NM_026731.3 Refseq
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 773044 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7xkh5:80/100.66.78.74:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0