This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01176661_g1
See other POLR2I GE Assays ›
Gene Symbol:
Polr2i
Gene Name:
polymerase (RNA) II (DNA directed) polypeptide I
Gene Aliases:
2810002B19Rik
Chromosome Location:
Chr.7: 30232074 - 30233387 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01176661_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Polr2i
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027259.1 NP_081535.1 2 - 3 385 65
GenBank mRNA AB221537.1 - 2 - 3 113 65
BC062812.1 - 2 - 3 385 3466417 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700000554.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase RNA metabolism protein nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

maintenance of transcriptional fidelity during DNA-templated transcription elongation from RNA polymerase II promoter
transcription-coupled nucleotide-excision repair
transcription, DNA-templated
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
nucleic acid binding
DNA binding
DNA-directed RNA polymerase activity
zinc ion binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700004731.mm.1 TC0700000554.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700004731.mm.1 NM_027259.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700004731.mm.1 NM_027259.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0