Assay Details
| Gene Symbol: Elavl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010487.2 | NP_034617.1 | 1 - 2 | 436 | 115 | |
| GenBank mRNA | BC052097.1 | - | 1 - 2 | 423 | 5695543 | 115 |
| U29148.1 | - | 1 - 2 | 415 | 115 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC052097.1, NM_010487.2, U29148.1 transcripts.
Set Membership: |
|

Elavl3
Genomic Map