This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01176859_g1
See other TMPRSS5 GE Assays ›
Gene Symbol:
Tmprss5
Gene Name:
transmembrane protease, serine 5 (spinesin)
Gene Aliases:
spinesin
Chromosome Location:
Chr.9: 49080439 - 49117591 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01176859_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmprss5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_030709.2 NP_109634.2 5 - 6 542 66
GenBank mRNA AB016229.1 - 5 - 6 540 66
AB016230.1 - 5 - 6 542 66
AB016423.1 - 1 - 2 137 66
AB041037.1 - 5 - 6 499 66
AK029714.1 - 6 - 7 1389 66
BC132151.1 - 4 - 5 488 40130428 66
BC145027.1 - 4 - 5 470 9053575 66

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AB016229.1, AB016423.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tmprss5

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
serine-type endopeptidase activity
scavenger receptor activity
peptidase activity
serine-type peptidase activity
hydrolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline