Assay Details
| Gene Symbol: Mpi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025837.2 | NP_080113.1 | 7 - 8 | 1087 | 84 | |
| GenBank mRNA | AF244360.1 | - | 7 - 8 | 1069 | 84 | |
| AK003289.1 | - | 5 - 6 | 847 | 84 | ||
| AK003896.1 | - | 7 - 8 | 1088 | 84 | ||
| AK132777.1 | - | 7 - 8 | 1088 | 84 | ||
| AK189289.1 | - | 2 - 3 | 236 | 84 | ||
| BC009068.1 | - | 7 - 8 | 1065 | 2650791 | 84 | |
| BC051163.1 | - | 7 - 8 | 1065 | 6466145 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542458 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542422 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542457 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542421 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P393897 | BC051163.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P393897 | AK132777.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P393897 | AK214803.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P393897 | AK218984.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P393897 | NM_025837.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P393897 | BC051163.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P393897 | AK132777.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P393897 | AK214803.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P393897 | AK218984.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P393897 | NM_025837.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 238847 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 607968 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 639359 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 81842 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 335538 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P158712 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P393897 | BC051163.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P393897 | AK132777.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P393897 | AK214803.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P393897 | AK218984.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P393897 | NM_025837.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P393897 | BC051163.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P393897 | AK132777.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P393897 | AK214803.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P393897 | AK218984.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P393897 | NM_025837.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P393897 | BC051163.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P393897 | AK132777.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P393897 | AK214803.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P393897 | AK218984.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P393897 | NM_025837.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P393897 | BC051163.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P393897 | AK132777.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P393897 | AK214803.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P393897 | AK218984.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P393897 | NM_025837.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542431 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542419 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542450 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542458 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542436 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542426 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542417 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542451 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542413 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542418 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542433 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542412 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542445 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542416 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542427 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542456 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542410 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542408 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542442 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542457 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542454 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542447 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542438 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542422 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542424 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542421 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542429 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542420 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26542453 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01941870 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P158712 | - | - |
| Whole Mouse Genome G4122A | A_51_P393897 | BC051163.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P393897 | AK132777.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P393897 | AK214803.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P393897 | AK218984.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P393897 | NM_025837.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P393897 | BC051163.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P393897 | AK132777.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P393897 | AK214803.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P393897 | AK218984.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P393897 | NM_025837.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P158712 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07675644 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07816032 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07595024 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07729366 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07816030 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07816031 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07706597 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07828976 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07747518 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542457 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542422 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542458 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26542421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26542304 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542410 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07590351 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542419 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07590352 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542414 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07845017 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542379 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08086867 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542420 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07845011 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542433 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542386 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542377 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542404 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542431 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08086863 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542405 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542399 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542371 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542427 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542426 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542391 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542380 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542422 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08110711 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07828978 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542368 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542397 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542367 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542400 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542383 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07887360 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542413 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542424 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542387 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542429 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542408 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08110709 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542394 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542417 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542418 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542369 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26542432 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07828970 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10593939 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593940 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593942 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593945 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593941 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593938 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593946 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593944 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593943 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451540_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10593937 | - | - |
| Mouse Genome 430 2.0 Array | 1451540_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1451540_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 110119_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 110119_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900019868.mm.1 | NM_025837.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900019868.mm.1 | NM_025837.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900019868.mm.1 | TC0900002374.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900019869.mm.1 | NM_025837.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900019869.mm.1 | NM_025837.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900019869.mm.1 | TC0900002374.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1451540_PM_at | - | - |
| Mouse Expression Set 430 | 1451540_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451540_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451540_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451540_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451540_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 559508 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2724022 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2724022 | - | - |

Mpi
Genomic Map