This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01177007_m1
See other PPP2R1B GE Assays ›
Gene Symbol:
Ppp2r1b
Gene Name:
protein phosphatase 2, regulatory subunit A, beta
Gene Aliases:
2410091N08Rik, AI790395
Chromosome Location:
Chr.9: 50856615 - 50899313 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01177007_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp2r1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001034085.2 NP_001029257.1 5 - 6 740 71
NM_001286553.1 NP_001273482.1 5 - 6 740 71
NM_028614.3 NP_082890.2 5 - 6 740 71
GenBank mRNA AK077158.1 - 5 - 6 694 71
AK136106.1 - 5 - 6 698 71
AK161392.1 - 5 - 6 693 71
BC056218.1 - 5 - 6 734 30016074 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900000609.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppp2r1b

Panther Classification:

Molecular Function -

phosphatase modulator

Gene Ontology Categories:

Function(s) Process(es)

apoptotic process involved in morphogenesis
positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900004372.mm.1 NM_001034085.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900004372.mm.1 NM_001286553.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900004372.mm.1 NM_028614.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900004372.mm.1 TC0900000609.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-pkm9c:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0