This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01177292_m1
See other DNMT3L GE Assays ›
Gene Symbol:
Dnmt3l
Gene Name:
DNA (cytosine-5-)-methyltransferase 3-like
Gene Aliases:
D6Ertd14e, ecat7
Chromosome Location:
Chr.10: 78042287 - 78063622 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01177292_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dnmt3l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081695.2 NP_001075164.1 7 - 8 722 66
NM_001284197.1 NP_001271126.1 5 - 6 563 66
NM_019448.4 NP_062321.1 6 - 7 576 66
GenBank mRNA AF220524.1 - 5 - 6 507 66
AJ404467.1 - 5 - 6 548 66
AK010434.1 - 6 - 7 577 66
BC083147.1 - 5 - 6 561 30447906 66
EF051624.1 - 7 - 8 722 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000000776.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dnmt3l

Panther Classification:

Molecular Function -

DNA methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
placenta development
methylation
positive regulation of catalytic activity
negative regulation of transcription, DNA-templated
chorionic trophoblast cell differentiation
genetic imprinting
protein binding
enzyme activator activity
enzyme binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000005687.mm.1 NM_001081695.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000005687.mm.1 NM_001284197.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000005687.mm.1 NM_019448.4 Refseq
Affymetrix Mouse Clariom S Array PSR1000005687.mm.1 TC1000000776.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0