This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01177293_m1
See other DNMT3L GE Assays ›
Gene Symbol:
Dnmt3l
Gene Name:
DNA (cytosine-5-)-methyltransferase 3-like
Gene Aliases:
D6Ertd14e, ecat7
Chromosome Location:
Chr.10: 78042287 - 78063622 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01177293_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dnmt3l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081695.2 NP_001075164.1 8 - 9 897 83
NM_001284197.1 NP_001271126.1 6 - 7 738 83
NM_019448.4 NP_062321.1 7 - 8 751 83
GenBank mRNA AF220524.1 - 6 - 7 682 83
AJ404467.1 - 6 - 7 723 83
AK010434.1 - 7 - 8 752 83
BC083147.1 - 6 - 7 736 30447906 83
EF051624.1 - 8 - 9 897 83

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dnmt3l

Panther Classification:

Molecular Function -

DNA methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
placenta development
methylation
positive regulation of catalytic activity
negative regulation of transcription, DNA-templated
chorionic trophoblast cell differentiation
genetic imprinting
protein binding
enzyme activator activity
enzyme binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rr9bx:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0