This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01177404_m1
See other NPAT GE Assays ›
Gene Symbol:
Npat
Gene Name:
nuclear protein in the AT region
Gene Aliases:
6820401K01, AI427561, BB112559
Chromosome Location:
Chr.9: 53537041 - 53575627 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01177404_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Npat
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081152.1 NP_001074621.1 10 - 11 1013 98
GenBank mRNA AK033012.1 - 10 - 11 993 98
AK080294.1 - 10 - 11 1014 98

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900000643.mm.1

Set Membership:

Amplicon length between 86 and 100 Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription involved in G1/S transition of mitotic cell cycle
transcription, DNA-templated
regulation of transcription, DNA-templated
cell cycle
regulation of gene expression
positive regulation of transcription, DNA-templated
negative regulation of nucleic acid-templated transcription
transcription coactivator activity
transcription corepressor activity
protein C-terminus binding
protein N-terminus binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900004592.mm.1 NM_001081152.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900004592.mm.1 NM_001081152.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900004592.mm.1 TC0900000643.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0