This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01179072_m1
See other SARS2 GE Assays ›
Gene Symbol:
Sars2
Gene Name:
seryl-aminoacyl-tRNA synthetase 2
Gene Aliases:
2410015F05Rik, D7Ertd353e, SerRS, SerRSmt
Chromosome Location:
Chr.7: 28741934 - 28753879 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01179072_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sars2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_023637.3 NP_076126.2 7 - 8 806 80
GenBank mRNA AB029949.1 - 7 - 8 778 80
AK010491.1 - 7 - 8 784 80
AK087606.1 - 7 - 8 773 80
AK140760.1 - 7 - 8 773 80
BC079664.1 - 7 - 8 783 30357834 80

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sars2

Panther Classification:

Molecular Function -

aminoacyl-tRNA synthetase translational protein

Gene Ontology Categories:

Function(s) Process(es)

translation
tRNA aminoacylation for protein translation
seryl-tRNA aminoacylation
nucleotide binding
aminoacyl-tRNA ligase activity
serine-tRNA ligase activity
ATP binding
ligase activity
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0