This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01179210_g1
See other TMEM25 GE Assays ›
Gene Symbol:
Tmem25
Gene Name:
transmembrane protein 25
Gene Aliases:
0610039J01Rik, AI429491
Chromosome Location:
Chr.9: 44793772 - 44799273 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01179210_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmem25
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027865.2 NP_082141.1 8 - 9 1087 91
GenBank mRNA AK002841.1 - 8 - 9 1088 91
AK154206.1 - 7 - 8 1573 91
AK165199.1 - 8 - 9 1119 91
AK165827.1 - 8 - 9 1072 91
BC060057.1 - 6 - 7 1410 5696102 91
BC060243.1 - 6 - 7 1410 5696102 91

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900002179.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tmem25

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26469618 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26469620 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26469619 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2126572 BC060243.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2126572 AK165827.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2126572 BC060057.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2126572 AK154206.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2126572 AK165199.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 145896 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 737472 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 889187 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P171266 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2126572 BC060243.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2126572 AK165827.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2126572 BC060057.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2126572 AK154206.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2126572 AK165199.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P142450 BC060243.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P142450 AK165827.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P142450 BC060057.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P142450 AK154206.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P142450 AK165199.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26469613 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26469608 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26469607 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26469611 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26469619 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26469618 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26469616 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P171266 - -
Whole Mouse Genome G4122A A_51_P142450 BC060243.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P142450 AK165827.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P142450 BC060057.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P142450 AK154206.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P142450 AK165199.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128421 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P171266 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07858827 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07858829 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07844440 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26469620 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26469618 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26469619 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26469651 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26469590 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26469597 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26469653 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26469649 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26469529 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26469595 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26469654 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26469607 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26469587 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26469618 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26469601 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26469643 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26469592 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26469605 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26469647 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26469535 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26469588 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26469534 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26469604 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26469613 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26469584 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26469646 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26469598 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26469586 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26469579 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26469619 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26469578 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26469611 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26469537 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26469580 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26469594 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26469531 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26469532 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26469616 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26469581 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26469605 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26469619 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26469595 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26469616 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26469597 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26469611 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08040092 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26469588 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08154349 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26469578 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26469579 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26469598 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08040063 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26469584 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07784390 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26469613 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26469580 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08040093 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26469604 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26469590 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26469594 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26469609 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26469592 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07858827 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10592929 - -
Affymetrix Mouse Gene 1.0 ST Array 10592931 - -
Affymetrix Mouse Gene 1.0 ST Array 10592927 - -
Affymetrix Mouse Gene 1.0 ST Array 10592934 - -
Affymetrix Mouse Gene 1.0 ST Array 10592937 - -
Affymetrix Mouse Gene 1.0 ST Array 10592928 - -
Affymetrix Mouse Gene 1.0 ST Array 10592930 - -
Affymetrix Mouse Gene 1.0 ST Array 10592933 - -
Affymetrix Mouse Gene 1.0 ST Array 10592932 - -
Affymetrix Mouse Gene 1.0 ST Array 10592935 - -
Affymetrix HT MG-430 PM Array Plate 1452962_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1436644_PM_x_at - -
Affymetrix Mouse Gene 1.1 ST Array 10592926 - -
Mouse Genome 430 2.0 Array 1452962_PM_at - -
Mouse Genome 430 2.0 Array 1452962_at - -
Mouse Genome 430 2.0 Array 1436644_PM_x_at - -
Mouse Genome 430 2.0 Array 1436644_x_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 71687_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 71687_PM_at - -
Affymetrix Mouse Clariom S Array PSR0900018120.mm.1 TC0900002179.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900018118.mm.1 NM_027865.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900018118.mm.1 NM_027865.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900018118.mm.1 TC0900002179.mm.1 MTA TC
Mouse Expression Set 430 1436644_x_at - -
Mouse Expression Set 430 1452962_at - -
Mouse Expression Set 430 1452962_PM_at - -
Mouse Expression Set 430 1436644_PM_x_at - -
Murine Genome U74v2 Set 165772_at - -
Murine Genome U74v2 Set 165772_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1452962_at - -
Affymetrix HT Mouse Genome 430B Array 1436644_PM_x_at - -
Affymetrix HT Mouse Genome 430B Array 1436644_x_at - -
Affymetrix HT Mouse Genome 430B Array 1452962_PM_at - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 4210129 - -
Illumina mouse-6 v1.0 expression beadchip 2570400 - -
Illumina mouseRef-8 v1.0 expression beadchip 0002570400 - -
Illumina mouseRef-8 v1.0 expression beadchip 0004210129 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2506074 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2495269 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2506074 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2495269 - -
mouse-6 v1.1 (Illumina) scl35940.9_671-S - -
mouse-6 v1.1 (Illumina) scl0003411.1_29-S - -
Illumina Sentrix mouse-6.v1 bead array 2570400 - -
Illumina Sentrix mouse-6.v1 bead array 4210129 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0003411.1_29-S - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl35940.9_671-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 2570400 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 4210129 - -
MouseRef-8 v2 Expression BeadChip ILMN_2506077 - -
MouseRef-8 v2 Expression BeadChip ILMN_2789294 - -
MouseRef-8 v2 Expression BeadChip ILMN_2506076 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2506077 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2789294 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2506076 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl35940.9_671-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline