This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01179271_m1
See other TMPRSS13 GE Assays ›
Gene Symbol:
Tmprss13
Gene Name:
transmembrane protease, serine 13
Gene Aliases:
BC010843, Tpmrss13
Chromosome Location:
Chr.9: 45319071 - 45347581 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01179271_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmprss13
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001013373.2 NP_001013391.2 10 - 11 1354 128
GenBank mRNA BC010843.1 - 2 - 3 203 128
BC042878.1 - 9 - 10 1090 128
BC085323.1 - 10 - 11 1339 30734093 128

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900000539.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tmprss13

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
establishment of skin barrier
molecular_function
serine-type endopeptidase activity
scavenger receptor activity
peptidase activity
serine-type peptidase activity
hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900003730.mm.1 NM_001013373.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900003730.mm.1 NM_001013373.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900003730.mm.1 TC0900000539.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900003729.mm.1 NM_001013373.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900003729.mm.1 NM_001013373.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900003729.mm.1 TC0900000539.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-dd86746ff-lmgjf:80/100.66.77.199:80.
git-commit: 04e44c58f20fafe66519586ffe6db5d8a90e52ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.53.1-Offline