This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01179524_m1
See other GLB1L2 GE Assays ›
Gene Symbol:
Glb1l2
Gene Name:
galactosidase, beta 1-like 2
Gene Aliases:
-
Chromosome Location:
Chr.9: 26763042 - 26806472 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01179524_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Glb1l2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_153803.1 NP_722498.1 11 - 12 1110 86
GenBank mRNA AK033400.1 - 7 - 8 693 86
AK136711.1 - 11 - 12 1144 86
AK143057.1 - 10 - 11 1041 86
BC038479.1 - 11 - 12 1110 4503030 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900001939.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Glb1l2

Panther Classification:

Molecular Function -

galactosidase

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
metabolic process
hydrolase activity, hydrolyzing O-glycosyl compounds
beta-galactosidase activity
lytic transglycosylase activity
mannosidase activity
mannosyl-oligosaccharide mannosidase activity
galactosidase activity
glucosidase activity
trehalase activity
fucosidase activity
hexosaminidase activity
amylase activity
hydrolase activity
hydrolase activity, acting on glycosyl bonds
dextrin alpha-glucosidase activity
starch alpha-glucosidase activity
beta-glucanase activity
beta-6-sulfate-N-acetylglucosaminidase activity
glucan endo-1,4-beta-glucosidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900016249.mm.1 NM_153803.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016249.mm.1 NM_153803.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016249.mm.1 TC0900001939.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900016250.mm.1 NM_153803.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016250.mm.1 NM_153803.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016250.mm.1 TC0900001939.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900016247.mm.1 NM_153803.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016247.mm.1 NM_153803.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016247.mm.1 TC0900001939.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900016248.mm.1 NM_153803.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016248.mm.1 NM_153803.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016248.mm.1 TC0900001939.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0