This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01179690_m1
See other SMPDL3A GE Assays ›
Gene Symbol:
Smpdl3a
Gene Name:
sphingomyelin phosphodiesterase, acid-like 3A
Gene Aliases:
0610010C24Rik, AI529588, ASM3A, ASML3, ASML3A
Chromosome Location:
Chr.10: 57794544 - 57811830 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01179690_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Smpdl3a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_020561.2 NP_065586.3 2 - 3 449 125
GenBank mRNA AK002455.1 - 2 - 3 388 125
AK151683.1 - 2 - 3 335 125
AK152282.1 - 2 - 3 350 125
AK205531.1 - 1 - 2 195 125
BC139288.1 - 2 - 3 341 8862310 125
BC139289.1 - 2 - 3 341 8862311 125
Y08135.1 - 2 - 3 403 125

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Smpdl3a

Panther Classification:

Molecular Function -

hydrolase phosphodiesterase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

sphingomyelin catabolic process
metabolic process
cyclic-nucleotide phosphodiesterase activity
3',5'-cyclic-AMP phosphodiesterase activity
calmodulin-dependent cyclic-nucleotide phosphodiesterase activity
cGMP-stimulated cyclic-nucleotide phosphodiesterase activity
cGMP-inhibited cyclic-nucleotide phosphodiesterase activity
photoreceptor cyclic-nucleotide phosphodiesterase activity
sphingomyelin phosphodiesterase activity
hydrolase activity
hydrolase activity, acting on glycosyl bonds
7,8-dihydro-D-neopterin 2',3'-cyclic phosphate phosphodiesterase activity
inositol phosphosphingolipid phospholipase activity
inositol phosphorylceramide phospholipase activity
mannosyl-inositol phosphorylceramide phospholipase activity
mannosyl-diinositol phosphorylceramide phospholipase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline