This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01180121_m1
See other SRRM1 GE Assays ›
Gene Symbol:
Srrm1
Gene Name:
serine/arginine repetitive matrix 1
Gene Aliases:
AA407769, POP101, Srm160
Chromosome Location:
Chr.4: 135320484 - 135353254 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01180121_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Srrm1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001130477.1 NP_001123949.1 3 - 4 325 98
NM_016799.3 NP_058079.2 3 - 4 325 98
GenBank mRNA AF062655.1 - 3 - 4 251 98
AK008284.1 - 3 - 4 253 98
AK028412.1 - 3 - 4 257 98
BC094322.1 - 3 - 4 240 6401250 98
BU514596.1 - 3 - 4 325 98

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400003760.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Srrm1

Panther Classification:

Molecular Function -

RNA metabolism protein RNA processing factor nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

RNA splicing, via transesterification reactions
mRNA splicing, via spliceosome
mRNA processing
RNA splicing
DNA binding
RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400030964.mm.1 TC0400003760.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400030964.mm.1 NM_001130477.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400030964.mm.1 NM_001130477.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400030964.mm.1 NM_016799.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400030964.mm.1 NM_016799.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wrkvw:80/100.66.78.186:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0