This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01180643_g1
See other PCMT1 GE Assays ›
Gene Symbol:
Pcmt1
Gene Name:
protein-L-isoaspartate (D-aspartate) O-methyltransferase 1
Gene Aliases:
C79501, PIMT
Chromosome Location:
Chr.10: 7629380 - 7680973 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01180643_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pcmt1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008786.2 NP_032812.2 5 - 6 874 157
GenBank mRNA AK006162.1 - 5 - 6 552 157
AK075632.1 - 4 - 5 361 157
AK148789.1 - 5 - 6 899 157
AK214928.1 - 2 - 3 141 157
BC040750.1 - 5 - 6 467 157
BC049613.1 - 5 - 6 578 157
BC058966.1 - 5 - 6 563 5717963 157
D38023.1 - 5 - 6 538 157
M60320.1 - 5 - 6 538 157

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000001698.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pcmt1

Panther Classification:

Molecular Function -

methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

cellular protein modification process
protein methylation
methylation
S-adenosylhomocysteine metabolic process
S-adenosylmethionine metabolic process
protein-L-isoaspartate (D-aspartate) O-methyltransferase activity
methyltransferase activity
S-adenosylmethionine-dependent methyltransferase activity
transferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000012405.mm.1 NM_008786.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000012405.mm.1 NM_008786.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000012405.mm.1 TC1000001698.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline