This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01180644_m1
See other PCMT1 GE Assays ›
Gene Symbol:
Pcmt1
Gene Name:
protein-L-isoaspartate (D-aspartate) O-methyltransferase 1
Gene Aliases:
C79501, PIMT
Chromosome Location:
Chr.10: 7629380 - 7680973 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01180644_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pcmt1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008786.2 NP_032812.2 6 - 7 957 61
GenBank mRNA AK006162.1 - 6 - 7 635 61
AK075632.1 - 5 - 6 444 61
AK208253.1 - 1 - 2 90 61
AK214928.1 - 3 - 4 224 61
BC040750.1 - 6 - 7 550 61
BC049613.1 - 6 - 7 661 61
BC058966.1 - 6 - 7 646 5717963 61
M60320.1 - 6 - 7 621 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000001698.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pcmt1

Panther Classification:

Molecular Function -

methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

cellular protein modification process
protein methylation
methylation
S-adenosylhomocysteine metabolic process
S-adenosylmethionine metabolic process
protein-L-isoaspartate (D-aspartate) O-methyltransferase activity
methyltransferase activity
S-adenosylmethionine-dependent methyltransferase activity
transferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000012402.mm.1 NM_008786.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000012402.mm.1 NM_008786.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000012402.mm.1 TC1000001698.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline