This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01181550_m1
See other ABCG2 GE Assays ›
Gene Symbol:
Abcg2
Gene Name:
ATP-binding cassette, sub-family G (WHITE), member 2
Gene Aliases:
ABC15, ABCP, AI428558, BCRP, Bcrp1, MXR, MXR1
Chromosome Location:
Chr.6: 58584491 - 58692451 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01181550_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Abcg2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011920.3 NP_036050.1 12 - 13 1949 132
GenBank mRNA AF140218.1 - 11 - 12 1512 132
BC053730.1 - 12 - 13 1961 30064535 132
KF662864.1 - 12 - 13 1546 132

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600000711.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Abcg2

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter primary active transporter transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
drug transmembrane transport
drug transport
urate metabolic process
drug export
embryonic process involved in female pregnancy
nucleotide binding
ATP binding
drug transmembrane transporter activity
ATPase activity
protein homodimerization activity
protein dimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600005460.mm.1 TC0600000711.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600005460.mm.1 NM_011920.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600005460.mm.1 NM_011920.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0