This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01181553_m1
See other ABCG2 GE Assays ›
Gene Symbol:
Abcg2
Gene Name:
ATP-binding cassette, sub-family G (WHITE), member 2
Gene Aliases:
ABC15, ABCP, AI428558, BCRP, Bcrp1, MXR, MXR1
Chromosome Location:
Chr.6: 58584491 - 58692451 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01181553_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Abcg2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK142528.1 - 1 - 2 79 71
GU597840.1 - 1 - 2 78 71
GU597841.1 - 1 - 2 78 71
GU597842.1 - 1 - 2 57 71
GU597844.1 - 1 - 2 31 71

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK142528.1, GU597840.1, GU597841.1, GU597842.1, GU597844.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter primary active transporter transporter

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0