This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01181599_m1
See other NDNF GE Assays ›
Gene Symbol:
Ndnf
Gene Name:
neuron-derived neurotrophic factor
Gene Aliases:
A930038C07Rik, epidermacan
Chromosome Location:
Chr.6: 65671611 - 65712329 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01181599_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ndnf
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172399.3 NP_765987.2 2 - 3 676 117
GenBank mRNA AK029210.1 - 2 - 3 677 117
BC047154.1 - 2 - 3 650 6489681 117

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600000765.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ndnf

Gene Ontology Categories:

Function(s) Process(es)

neuron migration
nervous system development
positive regulation of cell-substrate adhesion
positive regulation of neuron projection development
cell growth
peptide cross-linking via chondroitin 4-sulfate glycosaminoglycan
extracellular matrix organization
negative regulation of neuron apoptotic process
glycosaminoglycan binding
heparin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600005735.mm.1 TC0600000765.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600005735.mm.1 NM_172399.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600005735.mm.1 NM_172399.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rw9fd:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0