This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01181601_m1
See other SMARCAD1 GE Assays ›
Gene Symbol:
Smarcad1
Gene Name:
SWI/SNF-related, matrix-associated actin-dependent regulator of chromatin, subfamily a, containing DEAD/H box 1
Gene Aliases:
AV081750, AW226546, D6Pas1, Etl1, mKIAA1122
Chromosome Location:
Chr.6: 65042588 - 65116061 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01181601_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Smarcad1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001253392.1 NP_001240321.1 9 - 10 1440 80
NM_007958.1 NP_031984.1 11 - 12 1901 80
GenBank mRNA AK122454.1 - 11 - 12 1708 80
AK134442.1 - 11 - 12 1752 80
BC042442.1 - 9 - 10 1433 3980726 80
X69942.1 - 11 - 12 1901 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600000758.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Smarcad1

Panther Classification:

Molecular Function -

DNA helicase

Gene Ontology Categories:

Function(s) Process(es)

regulation of DNA recombination
DNA double-strand break processing
DNA repair
cellular response to DNA damage stimulus
chromatin modification
ATP-dependent chromatin remodeling
chromosome separation
histone H3 deacetylation
histone H4 deacetylation
nucleotide binding
DNA binding
helicase activity
ATP binding
hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600005678.mm.1 TC0600000758.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600005678.mm.1 NM_001253392.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600005678.mm.1 NM_001253392.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600005678.mm.1 NM_007958.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jmd5k:80/100.66.78.41:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0