This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01181930_g1
See other MAST2 GE Assays ›
Gene Symbol:
Mast2
Gene Name:
microtubule associated serine/threonine kinase 2
Gene Aliases:
MAST205, Mtssk
Chromosome Location:
Chr.4: 116306760 - 116464254 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01181930_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mast2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001042743.2 NP_001036208.1 23 - 24 3005 66
NM_008641.3 NP_032667.2 22 - 23 2984 66
GenBank mRNA AB093264.1 - 13 - 14 1637 66
AK004728.1 - 4 - 5 494 66
BC060703.1 - 23 - 24 2575 6412811 66
CF743521.1 - 3 - 4 342 66
U02313.1 - 22 - 23 2654 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400003337.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mast2

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
phosphorylation
regulation of interleukin-12 biosynthetic process
spermatid differentiation
nucleotide binding
magnesium ion binding
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
microtubule binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
phosphatase binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400026628.mm.1 TC0400003337.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400026628.mm.1 NM_001042743.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400026628.mm.1 NM_008641.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0