Assay Details
| Gene Symbol: Fabp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017399.4 | NP_059095.1 | 2 - 3 | 277 | 124 | |
| GenBank mRNA | AK131868.1 | - | 2 - 3 | 287 | 124 | |
| BC009812.1 | - | 2 - 3 | 277 | 4159971 | 124 | |
| Y14660.1 | - | 2 - 3 | 277 | 124 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fabp1
Genomic Map