This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01182545_m1
See other UROC1 GE Assays ›
Gene Symbol:
Uroc1
Gene Name:
urocanase domain containing 1
Gene Aliases:
AI265605, BC022133
Chromosome Location:
Chr.6: 90333239 - 90364551 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01182545_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Uroc1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_144940.2 NP_659189.2 2 - 3 289 57
GenBank mRNA AK149464.1 - 2 - 3 290 57
BC022133.1 - 2 - 3 272 5098655 57

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Uroc1

Panther Classification:

Molecular Function -

hydratase lyase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

histidine metabolic process
histidine catabolic process
urocanate hydratase activity
lyase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline