This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01182793_g1
See other CAPG GE Assays ›
Gene Symbol:
Capg
Gene Name:
capping protein (actin filament), gelsolin-like
Gene Aliases:
gCap39, mbh1
Chromosome Location:
Chr.6: 72544391 - 72562983 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01182793_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Capg
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001042534.3 NP_001035999.1 3 - 4 436 101
NM_001271395.1 NP_001258324.1 3 - 4 429 101
NM_001271415.1 NP_001258344.1 4 - 5 426 101
NM_007599.4 NP_031625.2 3 - 4 283 101
GenBank mRNA AK146622.1 - 3 - 4 287 101
AK149857.1 - 3 - 4 233 101
AK152818.1 - 3 - 4 261 101
AK159577.1 - 3 - 4 236 101
AK159736.1 - 3 - 4 238 101
AK165153.1 - 3 - 4 250 101
AK165586.1 - 1 - 2 79 101
BC003480.1 - 3 - 4 237 2811050 101
BU696808.1 - 3 - 4 283 101
CD551437.1 - 3 - 4 385 101
X54511.1 - 4 - 5 425 101

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Capg

Panther Classification:

Molecular Function -

non-motor actin binding protein

Gene Ontology Categories:

Function(s) Process(es)

cell projection assembly
barbed-end actin filament capping
actin filament capping
actin binding
protein domain specific binding
protein complex binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0