This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01183078_m1
See other SUMF1 GE Assays ›
Gene Symbol:
Sumf1
Gene Name:
sulfatase modifying factor 1
Gene Aliases:
AA543204, AI463102, AI851573, FGE
Chromosome Location:
Chr.6: 108107021 - 108185591 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01183078_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sumf1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145937.3 NP_666049.2 4 - 5 626 71
GenBank mRNA AK151874.1 - 4 - 5 627 71
AK159192.1 - 4 - 5 615 71
AK160917.1 - 4 - 5 608 71
AK161203.1 - 4 - 5 619 71
BC026981.1 - 4 - 5 599 5365580 71

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sumf1

Gene Ontology Categories:

Function(s) Process(es)

oxidation-reduction process
protein binding
dimethyl sulfoxide reductase activity
oxidoreductase activity
tetrachloro-p-hydroquinone reductive dehalogenase activity
protein homodimerization activity
metal ion binding
trichloro-p-hydroquinone reductive dehalogenase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0