This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01183873_m1
See other GFPT1 GE Assays ›
Gene Symbol:
Gfpt1
Gene Name:
glutamine fructose-6-phosphate transaminase 1
Gene Aliases:
2810423A18Rik, AI324119, AI449986, GFA, GFAT, GFAT1, GFAT1m, Gfpt
Chromosome Location:
Chr.6: 87042846 - 87092207 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01183873_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gfpt1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013528.3 NP_038556.1 17 - 18 1995 121
GenBank mRNA AF334736.1 - 18 - 19 2000 121
AK019852.1 - 17 - 18 1991 121
BC002283.1 - 14 - 15 1594 121
BC010516.1 - 17 - 18 1943 4191553 121
BC050762.1 - 17 - 18 1991 6742987 121
U00932.1 - 16 - 17 1842 121

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600001002.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gfpt1

Panther Classification:

Molecular Function -

transaminase

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
fructose 6-phosphate metabolic process
glucosamine biosynthetic process
UDP-N-acetylglucosamine metabolic process
glutamine metabolic process
metabolic process
circadian regulation of gene expression
negative regulation of glycogen biosynthetic process
rhythmic process
protein homotetramerization
carbohydrate derivative biosynthetic process
glutamine-fructose-6-phosphate transaminase (isomerizing) activity
transaminase activity
amino acid binding
transferase activity
carbohydrate binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600007868.mm.1 TC0600001002.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600007868.mm.1 NM_013528.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600007868.mm.1 NM_013528.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0