This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01183901_g1
See other CNBP GE Assays ›
Gene Symbol:
Cnbp
Gene Name:
cellular nucleic acid binding protein
Gene Aliases:
AA408710, Cnbp1, Znf9
Chromosome Location:
Chr.6: 87842615 - 87851106 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01183901_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cnbp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001109746.1 NP_001103216.1 3 - 4 416 61
GenBank mRNA AK075760.1 - 3 - 4 402 61
AK145444.1 - 3 - 4 367 61
AK146224.1 - 3 - 4 378 61
AK146301.1 - 3 - 4 359 61
AK146344.1 - 3 - 4 380 61
AK146367.1 - 3 - 4 364 61
AK146593.1 - 3 - 4 353 61
AK147157.1 - 3 - 4 344 61
AK151317.1 - 3 - 4 380 61
AK151528.1 - 3 - 4 350 61
AK151961.1 - 3 - 4 357 61
L12693.1 - 3 - 4 303 61
X63866.1 - 3 - 4 303 61
Z11870.1 - 3 - 4 303 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600002668.mm.1

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
positive regulation of cell proliferation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
nucleic acid binding
DNA binding
single-stranded DNA binding
single-stranded RNA binding
zinc ion binding
poly(A) RNA binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600020743.mm.1 TC0600002668.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600020743.mm.1 NM_001109745.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600020743.mm.1 NM_001109745.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600020743.mm.1 NM_001109746.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600020743.mm.1 NM_001109746.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600020743.mm.1 NM_013493.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600020743.mm.1 NM_013493.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0