This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01183914_m1
See other ARHGAP25 GE Assays ›
Gene Symbol:
Arhgap25
Gene Name:
Rho GTPase activating protein 25
Gene Aliases:
A130039I20Rik
Chromosome Location:
Chr.6: 87458544 - 87533259 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01183914_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Arhgap25
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001037727.2 NP_001032816.1 7 - 8 1253 69
NM_001286610.1 NP_001273539.1 6 - 7 1053 69
NM_175476.4 NP_780685.2 7 - 8 924 69
NR_104483.1 7 - 8 924 69
GenBank mRNA AK037710.1 - 7 - 8 986 69
AK048450.1 - 7 - 8 884 69
AK137444.1 - 7 - 8 912 69
AK137526.1 - 7 - 8 861 69
AK159598.1 - 6 - 7 1054 69
AK159941.1 - 6 - 7 1054 69
AK169897.1 - 7 - 8 1230 69
AK172878.1 - 5 - 6 583 69
BC108400.1 - 4 - 5 534 69
BC138751.1 - 6 - 7 832 8861773 69
BC138752.1 - 6 - 7 832 8861774 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600002656.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Arhgap25

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
molecular_function
GTPase activator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600020644.mm.1 TC0600002656.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600020644.mm.1 NM_001037727.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600020644.mm.1 NM_001286610.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600020644.mm.1 NM_175476.4 Refseq
Affymetrix Mouse Clariom S Array PSR0600020644.mm.1 NR_104483.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600020643.mm.1 TC0600002656.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600020643.mm.1 NR_104483.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0