This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01184342_m1
See other MBD4 GE Assays ›
Gene Symbol:
Mbd4
Gene Name:
methyl-CpG binding domain protein 4
Gene Aliases:
Med1
Chromosome Location:
Chr.6: 115840697 - 115853438 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01184342_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mbd4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010774.2 NP_034904.2 7 - 8 1715 86
GenBank mRNA AF072249.1 - 7 - 8 1732 86
AK147782.1 - 4 - 5 697 86
AK161005.1 - 7 - 8 2703 86
BC024812.1 - 7 - 8 1697 5356016 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600002963.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mbd4

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
base-excision repair
DNA methylation
cellular response to DNA damage stimulus
mitotic G2 DNA damage checkpoint
intrinsic apoptotic signaling pathway in response to DNA damage
response to radiation
purine-specific mismatch base pair DNA N-glycosylase activity
DNA binding
catalytic activity
uracil DNA N-glycosylase activity
pyrimidine-specific mismatch base pair DNA N-glycosylase activity
hydrolase activity
single-strand selective uracil DNA N-glycosylase activity
DNA N-glycosylase activity
DNA-3-methylbase glycosylase activity
nicotinamide riboside hydrolase activity
nicotinic acid riboside hydrolase activity
deoxyribonucleoside 5'-monophosphate N-glycosidase activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24773307 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24773309 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24773308 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P203192 BC024812.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P203192 AK161005.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P203192 AF072249.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P203192 AK147782.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P203192 NM_010774.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P203192 BC024812.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P203192 AK161005.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P203192 AF072249.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P203192 AK147782.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P203192 NM_010774.2 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 962433 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 716758 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 777195 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 212293 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 689574 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 297997 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 754821 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 84543 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P105029 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P130341 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P203192 BC024812.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P203192 AK161005.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P203192 AF072249.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P203192 AK147782.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P203192 NM_010774.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P203192 BC024812.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P203192 AK161005.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P203192 AF072249.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P203192 AK147782.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P203192 NM_010774.2 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P203192 BC024812.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P203192 AK161005.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P203192 AF072249.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P203192 AK147782.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P203192 NM_010774.2 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P203192 BC024812.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P203192 AK161005.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P203192 AF072249.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P203192 AK147782.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P203192 NM_010774.2 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24773315 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24773335 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24773321 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24773330 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24773322 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24773331 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24773314 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24773308 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24773320 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24773332 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24773323 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24773317 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24773309 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24773312 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24773310 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24773328 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24773319 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24773329 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06036972 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P105029 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P130341 - -
Whole Mouse Genome G4122A A_51_P203192 BC024812.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P203192 AK161005.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P203192 AF072249.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P203192 AK147782.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P203192 NM_010774.2 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P203192 BC024812.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P203192 AK161005.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P203192 AF072249.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P203192 AK147782.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P203192 NM_010774.2 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140678 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P130341 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P105029 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05093662 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05093654 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05093657 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05731984 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05731982 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24773307 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24773308 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24773309 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773308 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773234 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773204 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773199 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773198 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773338 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773355 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773223 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773212 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773344 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773210 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773208 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773215 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773347 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773203 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773336 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773225 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773321 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773328 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773227 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773231 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773193 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773354 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773213 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773315 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773310 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773361 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773357 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773333 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773236 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773309 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773221 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773202 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773356 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773218 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773189 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773330 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773349 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773319 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773226 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773353 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773207 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773239 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773329 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24773350 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24773336 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05093657 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24773317 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05343168 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24773321 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05731983 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24773356 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05375709 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24773325 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05434731 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24773308 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24773349 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24773236 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24773323 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24773333 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24773352 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24773328 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24773313 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24773234 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05609732 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24773239 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05499042 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05411393 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24773342 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24773341 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24773319 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05074062 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24773355 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05343165 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05325341 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24773353 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05343161 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24773344 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24773338 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24773345 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24773330 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24773357 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10547094 - -
Affymetrix Mouse Gene 1.0 ST Array 10547090 - -
Affymetrix Mouse Gene 1.0 ST Array 10547091 - -
Affymetrix Mouse Gene 1.0 ST Array 10547095 - -
Affymetrix Mouse Gene 1.0 ST Array 10547098 - -
Affymetrix Mouse Gene 1.0 ST Array 10547093 - -
Affymetrix Mouse Gene 1.0 ST Array 10547097 - -
Affymetrix Mouse Gene 1.0 ST Array 10547096 - -
Affymetrix Mouse Gene 1.0 ST Array 10547092 - -
Affymetrix Mouse Gene 1.0 ST Array 10547089 - -
Affymetrix HT MG-430 PM Array Plate 1449490_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1457708_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1441079_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10547088 - -
Mouse Genome 430 2.0 Array 1441079_PM_at - -
Mouse Genome 430 2.0 Array 1457708_PM_at - -
Mouse Genome 430 2.0 Array 1449490_PM_at - -
Mouse Genome 430 2.0 Array 1457708_at - -
Mouse Genome 430 2.0 Array 1449490_at - -
Mouse Genome 430 2.0 Array 1441079_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 17193_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 17193_at - -
Affymetrix Mouse Clariom S Array PSR0600022766.mm.1 TC0600002963.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600022766.mm.1 NM_010774.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600022766.mm.1 NM_010774.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600022771.mm.1 TC0600002963.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600022771.mm.1 NM_010774.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600022771.mm.1 NM_010774.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600022769.mm.1 TC0600002963.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600022769.mm.1 NM_010774.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600022769.mm.1 NM_010774.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600022770.mm.1 TC0600002963.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600022770.mm.1 NM_010774.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600022770.mm.1 NM_010774.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600022767.mm.1 TC0600002963.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600022767.mm.1 NM_010774.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600022767.mm.1 NM_010774.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600022765.mm.1 TC0600002963.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600022765.mm.1 NM_010774.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600022765.mm.1 NM_010774.2 Refseq
Mouse Expression Set 430 1449490_PM_at - -
Mouse Expression Set 430 1449490_at - -
Mouse Expression Set 430 1441079_at - -
Mouse Expression Set 430 1441079_PM_at - -
Mouse Expression Set 430 1457708_at - -
Mouse Expression Set 430 1457708_PM_at - -
Murine Genome U74v2 Set 103285_at - -
Murine Genome U74v2 Set 103285_PM_at - -
Mouse Genome 430A 2.0 Array 1449490_at - -
Mouse Genome 430A 2.0 Array 1449490_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1449490_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1449490_at - -
Affymetrix HT Mouse Genome 430B Array 1441079_at - -
Affymetrix HT Mouse Genome 430B Array 1441079_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1457708_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1457708_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 705007 - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 7100373 - -
Illumina mouseRef-8 v1.0 expression beadchip 0007100373 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1213981 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1213981 - -
mouse-6 v1.1 (Illumina) scl28518.8.1_30-S - -
Illumina Sentrix mouse-6.v1 bead array 7100373 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl28518.8.1_30-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 7100373 - -
MouseRef-8 v2 Expression BeadChip ILMN_1213981 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1213981 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl28518.8.1_30-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0