This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01184570_m1
See other OGG1 GE Assays ›
Gene Symbol:
Ogg1
Gene Name:
8-oxoguanine DNA-glycosylase 1
Gene Aliases:
Mmh
Chromosome Location:
Chr.6: 113326976 - 113334188 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01184570_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ogg1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010957.4 NP_035087.3 3 - 4 815 65
GenBank mRNA AF000669.1 - 3 - 4 693 65
AF003596.1 - 3 - 4 780 65
AK146870.1 - 3 - 4 815 65
AK167456.1 - 3 - 4 875 65
BC138733.1 - 3 - 4 771 8861755 65
BC138736.1 - 3 - 4 771 8861758 65
U88621.1 - 3 - 4 783 65
U96711.1 - 3 - 4 780 65
Y11247.1 - 3 - 4 746 65
Y13479.1 - 3 - 4 816 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600001246.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ogg1

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
base-excision repair
nucleotide-excision repair
regulation of transcription, DNA-templated
cellular response to DNA damage stimulus
response to oxidative stress
aging
metabolic process
response to radiation
response to estradiol
regulation of protein import into nucleus, translocation
nucleotide-excision repair, DNA incision
negative regulation of apoptotic process
nucleic acid phosphodiester bond hydrolysis
negative regulation of double-strand break repair via single-strand annealing
purine-specific mismatch base pair DNA N-glycosylase activity
oxidized base lesion DNA N-glycosylase activity
DNA binding
damaged DNA binding
catalytic activity
DNA-(apurinic or apyrimidinic site) lyase activity
uracil DNA N-glycosylase activity
microtubule binding
oxidized purine nucleobase lesion DNA N-glycosylase activity
hydrolase activity
hydrolase activity, acting on glycosyl bonds
lyase activity
single-strand selective uracil DNA N-glycosylase activity
DNA N-glycosylase activity
8-oxo-7,8-dihydroguanine DNA N-glycosylase activity
DNA-3-methylbase glycosylase activity
nicotinamide riboside hydrolase activity
nicotinic acid riboside hydrolase activity
deoxyribonucleoside 5'-monophosphate N-glycosidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600009648.mm.1 TC0600001246.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600009648.mm.1 NM_010957.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600009648.mm.1 NM_010957.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0