This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01184571_g1
See other OGG1 GE Assays ›
Gene Symbol:
Ogg1
Gene Name:
8-oxoguanine DNA-glycosylase 1
Gene Aliases:
Mmh
Chromosome Location:
Chr.6: 113326976 - 113334188 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01184571_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ogg1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010957.4 NP_035087.3 5 - 6 1149 89
GenBank mRNA AF000669.1 - 5 - 6 1027 89
AF003596.1 - 5 - 6 1114 89
AK008192.1 - 177 89
AK146870.1 - 5 - 6 1149 89
AK165875.1 - 5 - 6 1102 89
AK167456.1 - 5 - 6 1209 89
BC138733.1 - 5 - 6 1105 8861755 89
BC138736.1 - 5 - 6 1105 8861758 89
U88621.1 - 5 - 6 1117 89
U96711.1 - 5 - 6 1114 89
Y11247.1 - 5 - 6 1080 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600001246.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ogg1

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
base-excision repair
nucleotide-excision repair
regulation of transcription, DNA-templated
cellular response to DNA damage stimulus
response to oxidative stress
aging
metabolic process
response to radiation
response to estradiol
regulation of protein import into nucleus, translocation
nucleotide-excision repair, DNA incision
negative regulation of apoptotic process
nucleic acid phosphodiester bond hydrolysis
negative regulation of double-strand break repair via single-strand annealing
purine-specific mismatch base pair DNA N-glycosylase activity
oxidized base lesion DNA N-glycosylase activity
DNA binding
damaged DNA binding
catalytic activity
DNA-(apurinic or apyrimidinic site) lyase activity
uracil DNA N-glycosylase activity
microtubule binding
oxidized purine nucleobase lesion DNA N-glycosylase activity
hydrolase activity
hydrolase activity, acting on glycosyl bonds
lyase activity
single-strand selective uracil DNA N-glycosylase activity
DNA N-glycosylase activity
8-oxo-7,8-dihydroguanine DNA N-glycosylase activity
DNA-3-methylbase glycosylase activity
nicotinamide riboside hydrolase activity
nicotinic acid riboside hydrolase activity
deoxyribonucleoside 5'-monophosphate N-glycosidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600009651.mm.1 TC0600001246.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600009651.mm.1 NM_010957.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600009651.mm.1 NM_010957.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0