This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01184875_g1
See other MOGS GE Assays ›
Gene Symbol:
Mogs
Gene Name:
mannosyl-oligosaccharide glucosidase
Gene Aliases:
1810017N02Rik, AI181835, Gcs1
Chromosome Location:
Chr.6: 83115506 - 83118902 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01184875_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mogs
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_020619.2 NP_065644.2 1 - 2 419 90
GenBank mRNA AK144946.1 - 1 - 2 420 90
AK157286.1 - 1 - 2 416 90
BC051949.1 - 1 - 2 419 5716189 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600000922.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mogs

Panther Classification:

Molecular Function -

glucosidase

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
oligosaccharide metabolic process
molecular_function
catalytic activity
mannosyl-oligosaccharide glucosidase activity
hydrolase activity
hydrolase activity, acting on glycosyl bonds

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600006951.mm.1 NM_020619.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600006951.mm.1 NM_020619.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600006951.mm.1 TC0600000922.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600006950.mm.1 NM_020619.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600006950.mm.1 NM_020619.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600006950.mm.1 TC0600000922.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 393944 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline