This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01184876_g1
See other MOGS GE Assays ›
Gene Symbol:
Mogs
Gene Name:
mannosyl-oligosaccharide glucosidase
Gene Aliases:
1810017N02Rik, AI181835, Gcs1
Chromosome Location:
Chr.6: 83115506 - 83118902 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01184876_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mogs
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_020619.2 NP_065644.2 2 - 3 657 61
GenBank mRNA AK144946.1 - 2 - 3 658 61
AK157286.1 - 2 - 3 654 61
AK219249.1 - 1 - 2 126 61
BC051949.1 - 2 - 3 657 5716189 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600000922.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mogs

Panther Classification:

Molecular Function -

glucosidase

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
oligosaccharide metabolic process
molecular_function
catalytic activity
mannosyl-oligosaccharide glucosidase activity
hydrolase activity
hydrolase activity, acting on glycosyl bonds

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600006953.mm.1 NM_020619.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600006953.mm.1 NM_020619.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600006953.mm.1 TC0600000922.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline