This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01185113_m1
See other CD163 GE Assays ›
Gene Symbol:
Cd163
Gene Name:
CD163 antigen
Gene Aliases:
CD163v2, CD163v3
Chromosome Location:
Chr.6: 124304631 - 124330529 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01185113_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cd163
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001170395.1 NP_001163866.1 4 - 5 861 67
NM_053094.2 NP_444324.2 4 - 5 861 67
GenBank mRNA AF274883.1 - 4 - 5 856 67
BC144848.1 - 4 - 5 812 9053396 67
BC145793.1 - 4 - 5 812 40131096 67
DQ058616.1 - 4 - 5 817 67
DQ058617.1 - 4 - 5 817 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600001418.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cd163

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

receptor-mediated endocytosis
acute-phase response
inflammatory response
scavenger receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600011388.mm.1 TC0600001418.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600011388.mm.1 NM_001170395.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600011388.mm.1 NM_001170395.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600011388.mm.1 NM_053094.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600011388.mm.1 NM_053094.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-cj8gb:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline