This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01185277_g1
See other EXTL1 GE Assays ›
Gene Symbol:
Extl1
Gene Name:
exostoses (multiple)-like 1
Gene Aliases:
D430033M16Rik
Chromosome Location:
Chr.4: 134356373 - 134372547 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01185277_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Extl1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019578.2 NP_062524.2 2 - 3 1688 103
GenBank mRNA AF224461.1 - 2 - 3 920 103
AK085075.1 - 2 - 3 1690 103
AK161915.1 - 2 - 3 761 103
AK172619.1 - 2 - 3 1690 103
BC120890.1 - 2 - 3 920 40061413 103
BC120891.1 - 2 - 3 920 40061414 103

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400003738.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Extl1

Panther Classification:

Molecular Function -

glycosyltransferase

Gene Ontology Categories:

Function(s) Process(es)

glycosaminoglycan biosynthetic process
protein glycosylation
heparan sulfate proteoglycan biosynthetic process
transferase activity
transferase activity, transferring glycosyl groups
glucuronosyl-N-acetylglucosaminyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400030765.mm.1 TC0400003738.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400030765.mm.1 NM_019578.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400030765.mm.1 NM_019578.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hwq2n:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0